Skip to Content
MilliporeSigma

SML4320

Mizagliflozin

≥94% (HPLC)

Synonym(s):

3-[[3-[4-[[3-(β-D-Glucopyranosyloxy)-5-(1-methylethyl)-1H-pyrazol-4-yl]methyl]-3-methylphenoxy]propyl]amino]-2,2-dimethylpropanamide, GSK 1614235, KGA 3235

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H44N4O8
CAS Number:
Molecular Weight:
564.67
MDL number:
UNSPSC Code:
12352200
Assay:
≥94% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥94% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Mizagliflozin, an orally bioavailable antidiabetic agent and a highly selective human SGLT1 (hSGLT1) inhibitor (Kᵢ ≈ 27 nM, ~303-fold selectivity over SGLT2), competitively blocks glucose absorption in the small intestine, leading to its laxative effects. This inhibition increases luminal glucose concentration, creating an osmotic gradient that promotes water retention and triggers serotonin release, ultimately stimulating peristalsis and secretion, the key components of its laxative action. Furthermore, mizagliflozin has demonstrated the ability to improve vascular cognitive impairment by preventing hippocampal neuronal death, suggestive of blood-brain barrier permeability and potential central nervous system effects.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library