Skip to Content
MilliporeSigma

SML4323

Apatinib

≥98% (HPLC)

Synonym(s):

N-[4-(1-Cyanocyclopentyl)phenyl]-2-[(4-pyridinylmethyl)amino]-3-pyridinecarboxamide, N-[4-(1-cyanocyclopentyl)phenyl]-2-[(pyridin-4-ylmethyl)amino]pyridine-3-carboxamide, Rivoceranib, YN 968D1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H23N5O
CAS Number:
Molecular Weight:
397.47
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Apatinib (rivoceranib) is an orally available, potent, and selective inhibitor of vascular endothelial growth factor receptor 2 (VEGFR2) (IC50 = 1 nM) that potently blocks tumor angiogenesis and inhibits tumor growth. Apatinib exhibits encouraging antitumor efficacy and acceptable toxicities in multiple clinical trials. It also mildly targets c-Kit and c-SRC tyrosine kinases.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library