Skip to Content
MilliporeSigma

SML4324

VS-4718

≥98% (HPLC)

Synonym(s):

2-[[2-[[2-Methoxy-4-(4-morpholinyl)phenyl]amino]-5-(trifluoromethyl)-4-pyridinyl]amino]-N-methylbenzamide, 2-(2-(2-Methoxy-4-(4-morpholinyl)phenyl]amino)-5-(trifluoromethyl)pyridin-4ylamino)-N-methylbenzamide, PND-1186, PND 1186, PND1186, SR 2516, SR-2516, SR2516, VS 4718, VS4718

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H26F3N5O3
CAS Number:
Molecular Weight:
501.50
MDL number:
UNSPSC Code:
12352203
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

PND-1186 is an orally available, potent and reversible inhibitor against focal adhesion kinase (FAK IC50 = 1.5 nM; 50 µM ATP) and FMS-like tyrosine kinase 3 (72% Flt3 & 62% FAK inhibition by 100 nM PND-1186) with much reduced or little potenty toward 32 other kinases. PND-1186 inhibits cellular FAK activation (FAK pY397 IC50 = 100 nM) and migration (by 45%; 1 µM PND-1186) in adherent murine breast carcinoma 4T1 cultures. PND-1186 suppresses 4T1 soft agar colony growth (0.1-1 µM) and murine ID8 ovarian carcinoma tumor growth in mice in vivo (0.5 mg/mL via drinking water with 5% sucrose).


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library