Skip to Content
MilliporeSigma

SML4328

Mito-HE chloride

≥80% (HPLC)

Synonym(s):

Mito-hydroethidine chloride hydrochloride, [6-(3,8-diamino-6-phenyl-5(6H)-phenanthridinyl)hexyl]triphenylphosphonium chloride hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H43N3P·Cl · xHCl
Molecular Weight:
668.25 (free base basis)
UNSPSC Code:
12352200
Assay:
≥80% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥80% (HPLC)

form

powder

storage condition

desiccated

color

faint orange to red

solubility

DMSO: 2 mg/mL, clear

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

Mito-HE is a mitochondria superoxide probe that corresponds to dihydroethidium covalently linked to the mitochondria-targeting triphenylphosphonium (TPP+) cation. Mito-HE is widely used for live cell imaging. Cells are incubated with the probe at a final concentration of 0.3-5 μM for 10-30 min at 37°C before washing. Two different excitation wavelengths are used to distinguish the superoxide-dependent hydroxylation of Mito-HE (385-405 nm) from the nonspecific formation of ethidium (480-520 nm). DMSO stock solutions should be stored at -20°C for no more than 3 weeks. Please refer to Nat Protoc. 2008 3(6):941-7 (PMID 18536642) for experimental setup recommendations.

Disclaimer

Hygroscopic, store with desiccant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library