Skip to Content
MilliporeSigma

SML4330

MN714

≥98% (HPLC)

Synonym(s):

(S)-4-(3-((3-(2-Chloroacetamido)benzyl)amino)-2-(2-(4-fluorophenyl)acetamido)-3-oxopropyl)phenyl dipivaloyloxymethyl phosphate, MN 714, MN-714

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H46ClFN3O11P
Molecular Weight:
806.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

MN714 is a cell-permeable pivaloyloxymethyl (POM) ester precursor of MN551, an E3 ligase suppressor of cytokine signaling 2 (SOCS2) inhibitor. Upon entering cells, MN714 is hydrolyzed by cellular esterases to MN551 that covalently modifies SOCS2 SH2 domain EF loop Cys111. MN551 also effectively modifies SH2 containing protein (CISH) (Cys114), but not SOCS4 (Cys350) or SOCS6 (no EF loop Cys). MN714 treatment of K562 cells (10 µM for 6h) prior to lysate preparation compeletly abrogates endogenous SOCS2 binding of immobilized GHR pY595 substrate peptide.

Disclaimer

Hygroscopic, Store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library