Skip to Content
MilliporeSigma

SML4332

Lifitegrast sodium

≥98% (HPLC)

Synonym(s):

N-​[[2-​(6-Bbenzofuranylcarbonyl​)​-​5,​7-​dichloro-​1,​2,​3,​4-​tetrahydro-​6-​isoquinolinyl]​carbonyl]​-​3-​(methylsulfonyl)​-L-​phenylalanine sodium salt, SAR 1118 sodium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H23Cl2N2NaO7S
CAS Number:
Molecular Weight:
637.46
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Lifitegrast, a potent LFA-1 antagonist (IC50 ~2.98 nM), treats Dry Eye Disease by inhibiting T-cell mediated inflammation. It competitively and rapidly binds LFA-1 on T-lymphocytes, blocking interaction with ICAM-1, which inhibits crucial inflammatory processes like T-cell adhesion, migration, activation, and pro-inflammatory cytokine release (e.g., IFN-γ, TNF-α). This action downregulates T-cell activation signaling and inflammatory gene expression, producing targeted anti-inflammatory effects. Administered topically, Lifitegrast exhibits high ocular retention and bioavailability, ensuring localized biological action with minimal systemic exposure.

Disclaimer

Hygroscopic Store with desiccant


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library