Skip to Content
MilliporeSigma

SML4339

ASP2905

≥98% (HPLC)

Synonym(s):

ASP 2905, ASP2905, N2-(4-Fluorophenyl)-N4-phenyl-N6-(pyrimidin-2-ylmethyl)-1,3,5-triazine-2,4,6-triamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H17FN8
CAS Number:
Molecular Weight:
388.40
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

ASP2905, a cell-permeable, potent, selective and orally available KCNH3 inhibitor (IC50 = 9 nM in CHO cells expressing Kv12.2), regulates synthetic RNA riboswitches. Core action includes reversible, high-affinity binding to the AC17-4 RNA aptamer (KD = 7.7 nM), inducing a conformational change activating gene expression (>10-fold, at 5 µM) via ribozyme inhibition or exon skipping. In vivo, oral ASP2905 administration effectively induces reversible hEPO gene expression increase, notably in the liver of mice. Demonstrating low cytotoxicity, ASP2905 and AC17-4 provide a system for precise gene regulation, potentially controlling functional outcomes like myogenic differentiation via MyoD and apoptosis via BAX in HEK-293 cells.


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library