Skip to Content
MilliporeSigma

SML4344

Bitopertin

≥98% (HPLC)

Synonym(s):

RG1678, [4-(3-Fluoro-5-trifluoromethylpyridin-2-yl)piperazin-1-yl]-[5-methanesulfonyl-2-((S)-2,2,2-trifluoro-1-methylethoxy)phenyl]methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H20F7N3O4S
CAS Number:
Molecular Weight:
543.46
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Bitopertin (RG1678) is an orally available, potent (IC50=30nM) and selective inhibitor of glycine transporter GlyT1. Glycine transporters remove the neurtransmitter glycine from the synapse; their inhibition prolongs glycine transmission. GlyT1 inhibitors increase levels of glycine in the synaptic cleft leading to positive modulation of glutamatergic N-methyl-D-aspartate (NMDA) receptor function since glycine is a co-transmitter at NMDA receptors. Recent reports indicate that hypofunction of NMDA receptors may play an important role in the pathophysiology of schizophrenia, in particular the negative symptoms of the disease. Bitopertin is the first GlyT1 inhibitor to have a beneficial effect in schizophrenic patients.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library