Skip to Content
MilliporeSigma

SML4346

Norfloxacin hydrochloride

≥98% (HPLC)

Synonym(s):

1-Ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, MK-0366 HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H18FN3O3
CAS Number:
Molecular Weight:
319.33
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Norfloxacin hydrochloride (MK-0366 HCl) is a broad-spectrum fluoroquinolone antibiotic that is effective against both Gram-positive and Gram-negative bacteria. Norfloxacin is a potent inhibitor of bacterial DNA gyrase interaction with DNA.

Disclaimer

Hygroscopic Store with dessicant


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library