Skip to Content
MilliporeSigma

SML4348

ErSO-TFPy

≥98% (HPLC)

Synonym(s):

(R)-3-(4-Hydroxyphenyl)-3-(3,3,4,4-tetrafluoropyrrolidin-1-yl)-7-(trifluoromethyl)indolin-2-one, (3R)-1,3-dihydro-3-(4-hydroxyphenyl)-3-(3,3,4,4-tetrafluoro-1-pyrrolidinyl)-7-(trifluoromethyl)-2H-Indol-2-one, R-26

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H13F7N2O2
CAS Number:
Molecular Weight:
434.31
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

ErSO-TFPy potently inhibits multiple ERα+ breast cancer cell lines, exhibiting nanomolar IC50 values (≈ 5-25 nM). Its mechanism of action involves cation dysregulation, leading to cell swelling and the rapid induction of TRPM4-dependent necrotic cell death via activation of the anticipatory Unfolded Protein Response (a-UPR). Remarkably, a single dose of ErSO-TFPy achieves complete regressions of large tumors in preclinical models with sustained effect, despite its short half-life and transient blood exposure. This single-dose efficacy is coupled with excellent tolerability, demonstrated by high maximum tolerated doses (MTD) in mice (150 mg/kg IV), rats (>50 mg/kg IV), and dogs (>5 mg/kg IV), and a favorable therapeutic index, contrasting with multi-dose requirements of typical chemotherapies.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library