Skip to Content
MilliporeSigma

SML4349

LIH383 trifluoroacetate

≥95% (HPLC)

Synonym(s):

FGGFMRRK-NH2 trifluoroacetate, H-Phe-Gly-Gly-Phe-Met-Arg-Arg-Lys-NH2 trifluoroacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C45H72N16O8S · xC2HF3O2
Molecular Weight:
997.22 (free base basis)
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
(Powder or Lyophilized powder or film)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

LIH383 is a highly potent and highly selective agonist of scavenger receptor for chemokines ACKR3 (CXCR7) that potentiate opioid peptides activity towards classical receptors. It inhibits ACKR3’s negative regulatory function on opioid peptides in rat brain.

Disclaimer

Hygroscopic, Store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library