Skip to Content
MilliporeSigma

SML4350

UAMC-4821

≥98% (HPLC)

Synonym(s):

UAMC-4821 Ferroptosis inhibitor, UAMC4821, UAMC 4821

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H19N3O
CAS Number:
Molecular Weight:
257.33
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

UAMC-4821, a lipophilic radical-trapping antioxidant (RTA) and potent, orally bioavailable ferroptosis inhibitor, is designed to surpass the limitations of ferrostatin-1 and UAMC-3203. Demonstrating high potency (IC50 = 5.20 nM in HT-1080 cells, with FENIX assay confirming RTA activity), UAMC 4821 exhibits excellent oral bioavailability (63% in mice), low clearance, and high brain concentrations (123 ng/g after 24h, IV 10 mg/kg in mice), indicating superior ADME properties and blood-brain barrier penetration over earlier ferrostatins. Functionally, UAMC4821 directly neutralizes lipid peroxyl radicals within phospholipid bilayers, preventing membrane damage and ferroptotic death, thereby enhancing its attractiveness for CNS-targeted explorations.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library