Skip to Content
MilliporeSigma

SML4351

6PPD-Quinone

≥95% (HPLC)

Synonym(s):

6-PPD-Quinone, 6PPDQ, 6-PPDQ, 6PPD-Q

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H22N2O2
CAS Number:
Molecular Weight:
298.38
MDL number:
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

pink to dark red

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

6PPD-quinone (6PPDQ), a highly toxic tire antioxidant 6PPD derivative, bioaccumulates in lipid-rich tissues. 6PPD-Q primarily disrupts vascular permeability (potentially the blood-brain barrier) and induces mitochondrial dysfunction, causing oxidative stress. Molecularly, 6-PPDQ targets cannabinoid receptors (notably high-affinity CNR2), affects neuronal glycolysis, TCA intermediates, and promotes α-synuclein aggregation in vitro. Cellular/biological outcomes include dose-dependent mammalian multi-organ toxicity (hepatic, enteric, pulmonary, neuro-, reproductive [PCOS-linked], cardiac). In vivo, its extreme relevance is evident in potent salmonid lethality (e.g., Coho salmon LC50: 0.041 µg/L).


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library