Skip to Content
MilliporeSigma

SML4355

Ro 04-5595

≥95% (HPLC)

Synonym(s):

1-[2-(4-Chlorophenyl)ethyl]-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol, 1-(4-Chlorophenethyl)-6-methoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-7-ol, 1-[2-(4-Chlorophenyl)ethyl]-6-methoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-7-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H22ClNO2
CAS Number:
Molecular Weight:
331.84
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160702
Assay:
≥95% (HPLC)
Form:
oil
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

oil

color

colorless to yellow

storage temp.

2-8°C

Biochem/physiol Actions

Ro 04-5595 is a potent and selective allosteric antagonist for GluN2B (NR2B) containing NMDA receptors (Ki = 31 nM). Ro 04-5595 was predicted to bind to EVT-101 binding site, not the ifenprodil-binding site.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library