Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C12H15N5O3 · xHCl
CAS Number:
Molecular Weight:
277.28 (free base basis)
MDL number:
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥97% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
H2O: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Queuine is a hypermodified nucleobase found in the wobble anticodon position of tRNAs for amino acids His, Asn, Tyr, and Asp in most eukaryotes and prokaryotes. Queuine is a necessary coenzyme for eukaryotes supplied by diet and generated by symbiotic enteric bacteria.
Disclaimer
Hygroscopic, store with desiccant
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.