Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C26H17NO5
CAS Number:
Molecular Weight:
423.42
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
light yellow to orange
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Aminophenyl-fluorescein (APF), a cell-permeable and photostable probe, is designed for the selective detection of highly reactive oxygen and nitrogen species (hROS & hRNS), notably hypochlorite (OClˉ), hydroxyl radicals (HO•), and peroxynitrite (ONOOˉ) over minimally reactive H2O2, O2ˉ•, NO, 1O2 and ROO•. Initially exhibiting weak fluorescence (ε455 = 24,000 M-1cm-1, Φ: 0.008), APF undergoes O-dearylation upon reaction with these species, releasing its amino-phenoxy group and generating intensely fluorescent fluorescein, which leads to a dose-dependent fluorescence signal increase (λex/λem: 490/515 nm) and significantly enhanced optical properties (ε492 = 84,000 M-1cm-1, Φ: 0.85). APF′s inherent selectivity, complete autoxidation resistance, low toxicity, and quantitative fluorescence response establish it as a valuable tool for applications spanning microscopy, enzymatic assays, and multiplexing.
Disclaimer
Hygroscopic, store with desiccant
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
