Skip to Content
MilliporeSigma

SML4363

Tilmicosin phosphate salt

≥95% (HPLC)

Synonym(s):

4A-O-de(2,6-dideoxy-3-C-methyl-α-L-ribo-hexopyranosyl)-20-deoxo-20-[(3R,5S)-3,5-dimethyl-1-piperidinyl]-tylosin phosphate, Tilmicosin phosphate, LY 177370 phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H80N2O13·H3PO4
CAS Number:
Molecular Weight:
967.13
MDL number:
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Tilmicosin phosphate, a cell-permeable macrolide antibiotic with enhanced solubility and administration due to its phosphate salt, primarily targets bacterial 50S ribosomes, dose-dependently inhibiting protein synthesis. It is effective against Gram-positive bacteria, key Gram-negative respiratory pathogens like Pasteurella multocida (susceptible at ≤16 μg/ml) and Mannheimia haemolytica, and Mycoplasma species (e.g., M. gallisepticum MIC99 0.027 μg/mL), including notable activity against S. aureus (bovine mastitis MIC90 0.78 μg/mL versus native tilmicosin MIC 3.0 μg/mL), rendering it crucial for treating livestock respiratory diseases owing to its excellent lung penetration and prolonged half-life. Beyond selective antibacterial action, tilmicosin exhibits anti-inflammatory properties by modulating macrophage cytokine and gene expression (e.g., COX-2, iNOS).

Disclaimer

Hygroscopic, Store with desiccant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library