Skip to Content
MilliporeSigma

SML4365

GCN2-IN-1

≥98% (HPLC)

Synonym(s):

A 92, A-92, A92, GCN2 Inhibitor 1, SRI 41964, SRI-41964, SRI41964

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18N10O
CAS Number:
Molecular Weight:
402.41
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

GCN2-IN-1 (A92; A-92; SRI-41964) is a potent GCN2 inhibitor (IC50 <300 nM; 20 min inhibitor preincubation prior to 90 min kinase reaction at RT with 10 μM ATP & 80 nM eIF2α) that effectively downregulates GCN2 phosphorylation in U2OS cells (GCN2 pSer51 IC50 <300 nM post 2h treatment). A92 is also reported to inhibit the RNA-binding protein HuR dimerization (IC50 = 4.5 μM).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

68.0 °F

Flash Point (°C)

20 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library