Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C7H13NO4·HCl
CAS Number:
Molecular Weight:
211.64
UNSPSC Code:
12352203
eCl@ss:
32150313
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
H2O: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
SerBut (O-butyryl-L-serine) is an orally available, serine-conjugated butyrate precursor with therapeutic efficacy against autoimmune arthritis and neuroinflammation in vivo in mouse models of collagen antibody-induced arthritis (CAIA) (anti-collagen mAb on day 0, LPS i.p. on day 3, daily 20 mg SerBut/mouse p.o. starting on day -14 or twice daily 25 mg SerBut/mouse p.o. starting on day 4) and experimental autoimmune encephalomyelitis (EAE) (MOG35-55/CFA + PTX on day 0, daily 24 mg SerBut/mouse p.o. starting on day 2 with or twice daily 24 mg SerBut/mouse p.o. starting on day 2 without 100 mM SerBut in drinking water throughout starting day -14).
Disclaimer
Hygroscopic Store with desiccant
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.