Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C13H12N2O6· HCl
CAS Number:
Molecular Weight:
328.71
MDL number:
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
HADA, a non-toxic, blue-emitting (λex/λem: 405/450 nm) 7-hydroxycoumarin-labeled D-amino acid analog, covalently incorporates into bacterial peptidoglycan (PG) via periplasmic transpeptidases, primarily at D-alanine residues, enabling real-time visualization of PG dynamics, spatiotemporal growth, and cell wall synthesis across diverse bacteria. This highly versatile and specific live cell probe with moderate photostability facilitates multiplexed imaging and antibiotic and microbiome studies, utilizing bioorthogonal approaches for rapid, covalent PG labeling across a broad taxonomic range and permitting in situ studies.
Disclaimer
Hygroscopic, Store with desiccant
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.