Skip to Content
MilliporeSigma

SML4372

Adhibin

≥98% (HPLC)

Synonym(s):

3,6-Dibromo-1-hydroxymethyl-9H-carbazole, (3,6-Dibromo-9H-carbazol-1-yl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H9Br2NO
Molecular Weight:
355.02
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Adhibin is a selective allosteric inhibitor of RhoGAP class-IX myosins (Myo9a/b), with IC50 values of ~2.5 µM, disrupting ATPase and motor functions critical for RhoA/ROCK-mediated actin dynamics and contractility. It suppresses cancer cell adhesion, migration, and invasion in vitro, without significant toxicity up to 25 µM, and shows no systemic toxicity in vivo at doses up to 0.142 mg/kg (i.p.) in mice. Mechanistically, adhibin perturbs actomyosin structures, reduces active RhoA levels by up to 95%, and prevents focal adhesion formation, uniquely targeting metastasis-related processes. Additionally, adhibin induces cell rounding and detachment, delays cell cycle progression, and is effective in both single and collective migration contexts.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library