Skip to Content
MilliporeSigma

SML4374

HuR Molecular Glue Degrader, MG-HuR2

≥98% (HPLC)

Synonym(s):

Ethyl (E)-5-(4-(4-methoxyphenyl)-4-oxobut-2-enamido)-1H-indole-2-carboxylate, MG-HuR2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H20N2O5
CAS Number:
Molecular Weight:
392.40
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

light yellow to light orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

MG-HuR2 is a cell-permeable, druglike molecular glue that potently degrades the oncogenic RNA-binding protein HuR (DC₅₀ ≈ 5-10 nM in breast cancer cells) by recruiting the RNF126 E3 ligase. MG-HuR2’s strategic advantage lies in a novel dual-engagement mechanism, binding two distinct RNA-binding pockets (P1/P2) on HuR to drive a unique biphasic degradation profile, enabling more comprehensive and sustained target suppression than conventional inhibitors. This degradation cascade effectively downregulates key HuR-target mRNAs, reducing Bcl2 protein levels by up to 88% at a 10 nM concentration. Functionally, this translates to potent inhibition of cancer cell proliferation, induction of apoptosis, and the arrest of 3D spheroid growth. Given its favorable safety profile and precision mechanism, MG-HuR2 emerges as a powerful chemical tool for interrogating RNA biology and a promising therapeutic candidate for HuR-driven cancers.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library