Skip to Content
MilliporeSigma

SML4378

HSL119

≥98% (HPLC)

Synonym(s):

3-((8-Amino-1,7-naphthyridin-5-yl)ethynyl)-N-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H27F3N6O
CAS Number:
Molecular Weight:
544.57
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

HSL119 is a cell penetrant, potent and selective inhibitor of HUNK kinase (hormonally upregulated neu-associated kinase) that completely blocks phosphorylation of the Rubicon S92 in vitro. HSL119 decreases Rubicon S92 phosphorylation and potently inhibits survival of patient driven JIMT-1 cells (HER2+ breast cancer cells) (IC50 = 1.7µM in JIMT-1 cells). HSL119 exhibits substantial efficacy toward HER2+ breast cancer. Additionally, HSL119 inhibits ABL1, BRAF, FLT3, and RET kinases.


Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B - STOT RE 1 Dermal

Target Organs

Liver,Bone marrow,Pancreas,Cardio-vascular system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library