Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C13H8Cl2N2OS
CAS Number:
Molecular Weight:
311.19
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
-10 to -25°C
Biochem/physiol Actions
N6F11 is a ferroptosis inducer (0.5-10 µM for 24h) that causes cellular glutathione peroxidase 4 (GPX4) degradation (5 µM for 6-12h, 0.0625-5 µM for 12h) via ubiquitin-proteasome system (UPS) by inducing TRIM25-mediated ubiquitination. N6F11 causes ferroptotic cancer cell death that initiates HMGB1-dependent antitumor immunity mediated by CD8+ T-cells. When administered together with CD274 neutralizing antibody (clone 10F.9G2), N6F11 prolongs the average life expectancy of KPC mice, an orthotopic pancreatic ductal adenocarcinoma (PDA) model in vivo (10 mg N6F11/kg, 20 mg αCD274/kg via i.p.). Unlike RSL3, Fin56, and erastin, autophagy is dispensable of N6F11-induced ferroptosis.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
