Skip to Content
MilliporeSigma

SML4416

Tildipirosin

≥98% (HPLC)

Synonym(s):

20-Deoxo-23-deoxy-5-O-[3,6-dideoxy-3-(dimethylamino)-β-D-glucopyranosyl]-20,23-di-1-piperidinyltylonolide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C41H71N3O8
CAS Number:
Molecular Weight:
734.02
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Tildipirosin, a cell-permeable macrolide antibiotic, shows enhanced efficacy against key Gram-negative respiratory pathogens like Mannheimia haemolytica and Pasteurella multocida through improved outer membrane penetration. Selectively inhibits bacterial protein synthesis (IC50 ~0.23 µM) by binding the 23S rRNA of the 50S bacterial ribosomal subunit, blocking peptide chain prolongation and disrupting vital functions, yielding bacteriostatic or bactericidal effects with typical MIC90 values of 0.5-4 µg/ml. This mechanism enables effective treatment and metaphylaxis of Bovine (BRD) and Swine Respiratory Diseases (SRD), often as a single dose due to its long duration of action. Tildipirosin also displays immunomodulatory and anti-inflammatory actions by inhibiting pro-inflammatory cytokine secretion. Its significance is providing a targeted, long-acting therapeutic for critical veterinary respiratory infections.


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Repr. 2 - Skin Sens. 1 - STOT RE 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library