Skip to Content
MilliporeSigma

SML4417

USP30-I-1

≥98% (HPLC)

Synonym(s):

N-((3R,5R)-1-Cyano-5-methylpyrrolidin-3-yl)-5-(3-cyanophenyl)oxazole-2-carboxamide, USP30 Inhibitor-1, USP30-IN-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H15N5O2
CAS Number:
Molecular Weight:
321.33
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

USP30-I-1 is a potent and selective cyanopyrrolidine-based covalent inhibitor of the mitochondrial deubiquitinase USP30, designed to enhance mitophagy by targeting the catalytic cysteine (Cys77) with high specificity at concentrations ≤1 μM. USP30-I-1 demonstrates dose-dependent inhibition with an IC50 of 94 nM in neuroblastoma cells, efficiently blocking USP30 activity with no significant toxicity reported in the tested models and no adverse effects on non-target proteins within the tested concentration range. USP30-I-1’s kinetic profile (kinact/KI = 0.18 μM⁻¹s⁻¹) highlights its effective covalent bond formation, while in vitro studies confirm its ability to disrupt ubiquitin-substrate interactions essential for mitophagy regulation.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library