Skip to Content
MilliporeSigma

SML4419

ALKBH5 Inhibitor, Compound 20m

≥97% (HPLC)

Synonym(s):

5-Hydroxy-1-(3-(trifluoromethyl)phenyl)-1H-pyrazole-3-carboxylic acid, ALKBH5i-Compound 20m, Compound 20m

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H7F3N2O3
CAS Number:
Molecular Weight:
272.18
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Compound 20m potently inhibits RNA N⁶-methyladenosine (m⁶A) demethylase ALKBH5, with a fluorescence polarization assay IC₅₀ of 21 nM. Demonstrating high ALKBH5 selectivity, 20m shows minimal activity against FTO, ALKBH1, and ALKBH2 (IC₅₀ > 25 μM in DSF and FP assays). Cellular assays in HepG2 cells confirm 20m′s cell permeability and on-target demethylase activity, evidenced by ALKBH5 stabilization in cellular thermal shift assay and dose-dependent elevation of intracellular m⁶A levels. Molecular docking suggests inhibition occurs via 2-oxoglutarate binding pocket engagement, involving Mn(II) coordination, hydrogen bonds, and hydrophobic interactions.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library