Skip to Content
MilliporeSigma

SML4422

CAY10603

≥98% (HPLC)

Synonym(s):

BML 281, Isox, tert-Butyl (4-(3-((7-(hydroxyamino)-7-oxoheptyl)carbamoyl)isoxazol-5-yl)phenyl)carbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H30N4O6
CAS Number:
Molecular Weight:
446.50
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

CAY10603, also known as BML-281 or Isox, is a highly selective HDAC6 inhibitor featuring an active site zinc-binding isoxazolo hydroxamate chelator, with IC50 values of 2.4 nM for HDAC6, 71 nM for HDAC1, and ~ 500 nM for HDAC10. CAY 10603 demonstrates significant anticancer potential, especially against pancreatic cancer cells with IC50 values below 0.5 μM, while also inhibiting tau deacetylation on KXGS motifs and Hedgehog signaling via Gli-dependent and independent pathways. CAY-10603 exhibits potent anti-inflammatory effects, suppressing inflammatory cascades and oxidative stress by modulating NF-κB and NLRP3 inflammasome pathways in various conditions.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library