Skip to Content
MilliporeSigma

SML4443

Biotin-LC-Tyramide

≥98% (HPLC)

Synonym(s):

(3aS,4S,6aR)-Hexahydro-N-[6-[[2-(4-hydroxyphenyl)ethyl]amino]-6-oxohexyl]-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide, Biotin-LC-Phenol, Biotin-LC-Tyramine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H36N4O4S
CAS Number:
Molecular Weight:
476.63
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Biotin-LC-Tyramide is a bifunctional probe, featuring a phenol moiety for enzymatic oxidation and a biotin group for high-affinity streptavidin-based detection, engineered for high-fidelity proteomic mapping and signal amplification. Its core utility stems from an optimized 6-atom "LC" (Long Chain) aminohexanoic acid spacer, extending ~20.3 Å to position the biotin reporter away from the target protein surface, thereby mitigating steric hindrance for superior streptavidin binding. Activated by peroxidases, Biotin-LC-Tyramide generates a short-lived radical that covalently labels proximal tyrosine residues with high spatial resolution. This mechanism enables potent tyramide signal amplification (TSA) for high-resolution mapping in immunohistochemistry (IHC) and fluorescent in-situ hybridization and is a key reagent for proximity labeling techniques like APEX-MS to identify organelle-specific proteomes and protein-protein interactions in living cells.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library