Skip to Content
MilliporeSigma

SML4450

S-2238 acetate

≥98% (HPLC)

Synonym(s):

D-Phe-Pip-Arg-pNA acetate, D-Phe-Piperidinecarbonyl-Arg-p-nitroanilide acetate, D-Phenylalanyl-L-2-piperidinecarbonyl-N-(4-nitrophenyl)-L-argininamide acetate, H-D-Phe-Pip-Arg-pNA acetate, S 2238 acetate, S2238 acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H36N8O5
Molecular Weight:
552.63
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

storage temp.

-10 to -25°C

Biochem/physiol Actions

S-2238 is a chromogenic (colorimetric) thrombin substrate. Thrombin cleavage resluts in the release of the c-terminal p-nitroaniline (pNA; absorbance max 395 +/- 10 nm).

Disclaimer

Hygroscopic, Store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library