Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C16H14O6
CAS Number:
Molecular Weight:
302.28
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
light yellow to orange
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
Hematoxylin, a natural polyphenolic compound, is oxidized to its active form, hematein. This hematein functions as a chelator that, when complexed with a metal mordant (typically Al³⁺ or Fe³⁺), forms a cationic dye complex. This complex serves as the definitive acidic nuclear stain by electrostatically binding to negatively charged targets, primarily the phosphate groups of DNA and acidic nuclear proteins like histones. This mechanism confers high selectivity for the nucleus over the cytoplasm and ensures excellent retention in fixed and permeabilized tissues. As the gold standard in histopathology, it provides unparalleled, permanent nuclear counterstaining. When paired with eosin (H&E), it yields exceptionally high contrast—clearly demarcating the blue/purple nucleus from the pink/red cytoplasm—making it indispensable for providing morphological context in techniques like IHC. Beyond this core utility, hematoxylin serves specialized roles, including the effective staining of sperm cells for forensic analysis and the inhibition of amyloid β-protein fibrillation. Furthermore, an iron-hematein chelate exhibits NIR emission, enabling its use as a natural biosensor for detecting oxidative stress.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
