Skip to Content
MilliporeSigma

SML4453

MYTsA, Coupling Reagent

≥98% (HPLC)

Synonym(s):

MYTsA, Ynamide, N-Ethynyl-N,4-dimethylbenzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H11NO2S
CAS Number:
Molecular Weight:
209.26
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Ynamide, MYTsA is a bench-stable reagent that activates carboxylic acids via α-acyloxyenamide formation, enabling rapid, high-yield amide and peptide bond formation. MYTsA couples free carboxyl and amino groups without racemization/epimerization, even in the presence of common protecting groups and diverse side-chain functionalities such as −OH, −SH, −CONH₂, and the NH of indole. The reaction accelerates in polar solvents, with water serving as an eco-friendly cosolvent. MYTsA facilitates peptide cyclization, esterification, macrolactonization, thioamide/thioester formation, and inverse peptide synthesis via a transient protection strategy, while promoting the multicomponent assembly of β-amino amides from aldehydes, anilines, and carboxylic acids.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library