Skip to Content
MilliporeSigma

SML4454

Elacestrant dihydrochloride

≥98% (HPLC)

Synonym(s):

(6R)-6-[2-[Ethyl[[4-[2-(ethylamino)ethyl]phenyl]methyl]amino]-4-methoxyphenyl]-5,6,7,8-tetrahydro-2-naphthalenol hydrochloride, RAD 901 dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H38N2O2·2HCl
CAS Number:
Molecular Weight:
531.56
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Elacestrant (RAD1901) is an orally bioavailable, non-steroidal selective estrogen receptor degrader (SERD) and ER antagonist that potently inhibits ERα-mediated proliferation (e.g., 4 pM EC50 in MCF-7 cells, 48 nM ERα IC50) while showing minimal activity against ERβ (870 nM IC50). Elacestrant induces dose-dependent ERα degradation via the proteasome, a dual mechanism distinct from PI3K/AKT/mTOR pathways, conferring efficacy in preclinical models resistant to fulvestrant and CDK4/6 inhibitors, including those with ESR1 mutations. Elacestrant also penetrates the blood-brain barrier and shows synergistic potential with palbociclib or everolimus, positioning it as a key agent for advanced ER+ breast cancer.

Disclaimer

Hygroscopic, store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library