Skip to Content
MilliporeSigma

SML4455

Y-27632 2HCl ≥ 99% by HPLC

≥99% (HPLC)

Synonym(s):

ROCK Inhibitor, Rho-Associated Kinases Inhibitor, trans-4-[(R)-1-Aminoethyl]-N-(4-pyridyl)cyclohexanecarboxamide dihydrochlorid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H21N3O·2HCl
CAS Number:
Molecular Weight:
320.26
UNSPSC Code:
12352200
Assay:
≥99% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Y-27632 is a reversible, cell-permeable, ATP-competitive inhibitor of ROCK-I and ROCK-II (Ki~0.2–0.3 μM) that targets the catalytic cleft to fundamentally modulate cytoskeletal mechanics. By blocking actomyosin contractility, it induces the disassembly of actin stress fibers and focal adhesions, preventing membrane blebbing and driving potent smooth muscle relaxation (IC50~0.3–1.0 μM). While effective in hypertensive models, its systemic utility is constrained by rapid clearance and the risk of profound hypotension, often necessitating local administration. Consequently, its therapeutic potential is best realized in specialized tissues: Y27632 crosses the blood-brain barrier to promote axon regeneration and prevent growth cone collapse in CNS injury, and in ophthalmology, it lowers intraocular pressure and aids corneal endothelial healing. Beyond these in vivo applications, Y-27632 is the industry standard for ex vivo cell manipulation, particularly in cancer biology to inhibit metastasis and, most significantly, in stem cell research. Here, Y-27632 prevents dissociation-induced apoptosis (anoikis) in human pluripotent stem cells (hESCs/iPSCs), revolutionizing single-cell cloning, enzymatic passaging, and graft viability in regenerative medicine.

Disclaimer

Hygroscopic, Store with desiccant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library