Skip to Content
MilliporeSigma

SML4456

Isoquercetin ≥ 98% by HPLC

≥98% (HPLC)

Synonym(s):

2-(3,4-Dihydroxyphenyl)-3-(β-D-glucopyranosyloxy)-5,7-dihydroxy-4H-1-benzopyran-4-one; 3-Glucosylquercetin, 3′,4′,5,7-Tetrahydroxyflavone-3-β-D-glucopyranoside, Quercetin 3-O-β-D-glucopyranoside, Quercetin-3-O-glucoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Isoquercetin (Quercetin-3-O-glucoside) functions as a highly orally bioavailable "pro-drug" for quercetin, leveraging SGLT1-mediated active transport for rapid intestinal absorption. Following uptake, it undergoes immediate deglycosylation and extensive phase II metabolism—including glucuronidation, sulfation, and methylation—within enterocytes and the liver, circulating primarily as conjugated metabolites. Mechanistically, Isoquercetin delivers potent pleiotropic efficacy: it inhibits oncogenic Wnt/β-catenin signaling, blocks viral replication via SARS-CoV-2 3CLpro and Influenza A PB2 (ED50 1.2 µM) inhibition, and exerts significant antithrombotic activity by inhibiting Protein Disulfide Isomerase (PDI; IC50 2.5 μM) and blocking platelet factor Va/thrombin generation (IC50 ~5 μM). Additionally, it restores redox homeostasis through Nrf2/HO-1 induction and suppresses neuroinflammation via NF-κB downregulation and BDNF upregulation. Distinguished by superior pharmacokinetics over quercetin and a GRAS-certified safety profile, Isoquercetin represents a robust broad-spectrum therapeutic for metabolic, infectious, thrombotic, and neurodegenerative pathologies.

Disclaimer

Hygroscopic, Store with desiccant


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library