Skip to Content
MilliporeSigma

SML4458

Lithocholic acid High Purity

≥98% (HPLC)

Synonym(s):

3α-Hydroxycholanic acid; (3α,5β)-3-Hydroxycholan-24-oic acid, 17β-(1-Methyl-3-carboxypropyl)etiocholan-3α-ol; 3α-Hydroxy-5β-cholan-24-oate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H40O3
CAS Number:
Molecular Weight:
376.57
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Lithocholic acid (LCA), a secondary bile acid absorbed into the bloodstream and transported to muscle, acts as a TGR5 agonist and endogenous calorie restriction mediator, indirectly activating SIRT1 via TUB Like Protein 3 (TULP3) to engage AMPK through v-ATPase inhibition. This cascade improves metabolic parameters and stress resistance, raises GLP-1, and notably exhibits antineoplastic effects in pancreatic adenocarcinoma by inducing oxidative/nitrosative stress via CAR, FXR, and VDR nuclear receptors, inhibiting proliferation, EMT, and stem cell markers while upregulating mitochondrial oxidative phosphorylation. LCA extends lifespan in model organisms and enhances grip strength and insulin sensitivity in aged mice, underscoring its broad therapeutic potential in aging and oncology.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library