Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C19H18CuN4O2S2
CAS Number:
Molecular Weight:
462.05
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
faint orange to red
solubility
H2O: 2 mg/mL, clear (Warmed)
Biochem/physiol Actions
Elesclomol-Cu(II), is a cell-permeable, complex of potent copper ionophore Elesclomol with copper II (Cu2+) that selectively targets the mitochondria of cancer cells reliant on oxidative phosphorylation (OXPHOS). Its core action involves ferredoxin 1 (FDX1)-mediated reduction within the mitochondrial matrix, releasing toxic Cu(I) to initiate a dual cell death cascade. This comprises both overwhelming oxidative stress leading to apoptosis and cuproptosis, a distinct, genetically-defined mechanism driven by the aggregation of lipoylated TCA cycle proteins, whose core components (such as FDX1 and DLAT) serve as key biomarkers. This unique FDX1-dependent process confers nanomolar cytotoxicity and synergistic in vivo efficacy, making it an invaluable probe for dissecting the interplay between metal homeostasis, mitochondrial metabolism, and proteotoxic stress.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

