Skip to Content
MilliporeSigma

SML4460

Ph_N3, Clickable APEX2 Probe

≥98% (HPLC)

Synonym(s):

1-Azido-2-(4-hydroxyphenyl)ethane, 4-(2-Azidoethyl)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H9N3O
CAS Number:
Molecular Weight:
163.18
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
oil
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

oil

color

colorless to yellow

storage temp.

-10 to -25°C

Biochem/physiol Actions

Ph_N3 is a cell-permeable, clickable phenol probe designed for high-efficiency APEX2-catalyzed RNA proximity labeling in live cells, where a brief H₂O₂ pulse generates a short-lived phenoxyl radical for irreversible covalent tagging of proximal biomolecules within a ~20 nm radius. This mechanism provides superior performance over biotin-phenol for spatially resolved RNA mapping (APEX-seq), demonstrated by the high-confidence enrichment of 15 mitochondrial transcripts in mito-APEX2 cells with minimal organelle perturbation. Ph_N3’s clickable azide handle offers dual bioorthogonal routes (CuAAC/SPAAC) for versatile downstream analysis. Critically, its mechanistic orthogonality to singlet oxygen (¹O₂)-mediated chemistry is a pivotal advantage, enabling unprecedented sequential dual-location labeling protocols to investigate biomolecular dynamics.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library