Skip to Content
MilliporeSigma

SML4461

Alkyne-phenol-II

≥98 (HPLC)

Synonym(s):

Alk-Ph-II, Alk-Phenol-II, Alkyne tyramide-II, N-[2-(4-Hydroxyphenyl)ethyl]-4-pentynamide, Ph_Amide_Yne, clickable APEX probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H15NO2
CAS Number:
Molecular Weight:
217.26
UNSPSC Code:
12352200
Assay:
≥98 (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98 (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Alkyne tyramide-II (Alkyne-phenol-II, Alk-Ph-II, Ph_Amide_Yne) is a clickable alkyne-phenol substrate engineered to overcome the yeast cell wall permeability barrier that renders standard biotin-phenol (BP) probes ineffective. By replacing the bulky biotin with a small, highly soluble alkyne handle, Alk-Ph-II enables efficient labeling in intact cells, circumventing the need for artifact-inducing pretreatments. Upon H₂O₂ activation, genetically targeted APEX2 catalyzes the one-electron oxidation of Alk-Ph-II, generating a short-lived phenoxyl radical. This intermediate irreversibly tags proximal proteins and RNA within a ~10 nm radius in a rapid, 1-minute reaction, which is then captured via CuAAC click chemistry. This strategy delivers high-specificity (94%) proteomic maps, identifying 165 mitochondrial matrix proteins, and uniquely facilitates the first successful proximity-dependent transcriptome profiling (APEX-seq) in yeast.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library