Skip to Content
MilliporeSigma

SML4469

BODIPY FL-C3

≥98% (HPLC)

Synonym(s):

3-Bodipy-propanoic Acid, BODIPY 503/512, Bodipy FL, Bodipy FL 510, Hydrogen (T-4)-[5-[(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)methyl]-1H-pyrrole-2-propanoato(2-)-κN1]difluoroborate(1-)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H14BF2N2O2·H
CAS Number:
Molecular Weight:
292.09
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

faint orange to red

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

BODIPY FL C3 carboxylic acid is a high-performance labeling reagent distinguished by its core BODIPY fluorophore, which provides exceptionally bright (ε ≈ 85,000 M⁻¹cm⁻¹, Φ > 0.9), photostable, and sharp green fluorescence (λex/λem ~503/509-512 nm). Bodipy FL’s key strategic advantage is the remarkable stability of its fluorescence across wide variations in environmental pH and polarity, overcoming a primary limitation of traditional dyes. The integrated C3 carboxylic acid linker serves as a versatile chemical handle, enabling not only direct covalent attachment to primary amines on biomolecules but also serving as a key precursor for synthesizing advanced tools, such as clickable biorthogonal probes. This versatility yields intensely fluorescent and highly stable conjugates ideal for quantitative microscopy, flow cytometry, and other demanding bio-imaging applications.

Disclaimer

Hygroscopic, Store with desiccant


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library