Skip to Content
MilliporeSigma

SML4473

EL133, Keap1 PINCH

≥96% (HPLC)

Synonym(s):

N28,N28′-[1,2-ethanediylbis(oxy-2,1-ethanediyl)]bis[2-cyano-3,12-dioxo-Oleana-1,9(11)-dien-28- amide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C68H94N4O8
CAS Number:
Molecular Weight:
1095.50
UNSPSC Code:
12352200
Assay:
≥96% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥96% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

EL133 is a highly cell-permeable Polymerization-Inducing Chimera (PINCH) rationally designed to exploit protein symmetry (homodimerism) as a selective vulnerability for functional protein knockout. Comprising two bardoxolone (BDX) moieties conjugated via a precise short polyethylene glycol (PEG2) linker, EL-133 acts as a covalent molecular bridge targeting Cysteine 151 (C151) within the Keap1 BTB domain to induce supramolecular assembly. This mechanism drives the rapid (>90%) and high-specificity precipitation of Keap1 from the soluble cytosolic fraction (insolubility EC50 = 102 nM in OCI-AML2; effective >500 nM), with global proteomics confirming COL1A1 as the sole significant off-target. By locking Keap1 in an inactive polymeric state, EL133 triggers robust, proteasome-independent activation of the Nrf2 pathway and Antioxidant Response Element (ARE) genes, resulting in distinct, sustained upregulation of NAD(P)H quinone dehydrogenase 1 (NQO1). Distinct from PROTACs or simple inhibitors, EL-133 functions as an autonomous sequestrator independent of cellular degradation machinery (lysosome/proteasome) or E3 ligase co-expression, effectively circumventing resistance mechanisms associated with effector dependency.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library