Skip to Content
MilliporeSigma

SML4475

Ponatinib hydrochloride

≥98% (HPLC)

Synonym(s):

3-(Imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)benzamide Hydrochloride, AP24534 HCl, Ponatinib HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H27F3N6O · xHCl
Molecular Weight:
532.56 (free base basis)
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

AP24534 (Ponatinib) is a reversible, ATP-competitive, BBB-permeable and orally active pan-BCR-ABL inhibitor engineered to overcome multidrug-resistant CML. Its design features a rigid ethynyl linker that bypasses the steric hindrance of the T315I "gatekeeper" mutation, yielding profound potency against native ABL (IC50 0.37 nM), T315I (IC50 2.0 nM), and T315I-inclusive compound mutants (IC50 0.30–0.44 nM). This efficacy extends to cellular models, where it suppresses CrkL phosphorylation and inhibits proliferation in native (IC50 0.5 nM) and T315I-expressing (IC50 11 nM) cells. Beyond ABL, Ponatinib acts as a multi-kinase inhibitor modulating angiogenesis and stromal signaling, with high affinity for LYN (0.24 nM), PDGFRα (1.1 nM), VEGFR2 (1.5 nM), FGFR1-4 (~2.2 nM), SRC (5.4 nM), KIT (~12.5 nM), and FLT3 (~13 nM). Mechanistically, it halts STAT5, ERK, and AKT signaling to trigger cell cycle arrest and intrinsic apoptosis—driven by caspase-3 cleavage and Bcl-xL/Mcl-1 downregulation—or RIPK1/3-mediated necroptosis. To deepen molecular response against complex resistance, Ponatinib demonstrates significant synergy with MDM2 inhibitors, the allosteric inhibitor asciminib, and the BCL-2 inhibitor venetoclax.

Disclaimer

Hygroscopic, Store with desiccant


Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - STOT RE 1 Dermal

Target Organs

Liver,Bone marrow,Pancreas,Cardio-vascular system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Codes



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library