Skip to Content
MilliporeSigma

SML4476

Tradipitant

≥98% (HPLC)

Synonym(s):

LY-686017, LY686017, VLY-686, [2-[1-[[3,5-bis(Trifluoromethyl)phenyl]methyl]-5-pyridin-4-yltriazol-4-yl]pyridin-3-yl]-(2-chlorophenyl)methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H16ClF6N5O
CAS Number:
Molecular Weight:
587.90
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Tradipitant (LY686017) is a non-peptidyl tachykinin receptor 1 (TACR1/NK1R) antagonist optimized for superior metabolic stability and high oral bioavailability. Mechanistically, it functions via competitive antagonism of the preferred endogenous ligand, Substance P (SP), binding within the receptor’s transmembrane core to sterically hinder the recruitment of G-proteins and β-arrestins. This molecular blockade abolishes Gq/11-coupled signaling, thereby preventing phospholipase C activation, IP3/DAG release, intracellular calcium mobilization, and downstream MAPK/ERK phosphorylation cascades. Characterized by reversible binding with slow dissociation kinetics in striatal tissue that ensures sustained receptor occupancy, tradipitant demonstrates a Kd of 0.34 nM and a Bmax of 31.4 fmol/mg in guinea pig striatum. While highly selective for NK1R over NK2 and NK3 subtypes, it exhibits critical species-selectivity, possessing high affinity for human, guinea pig, and gerbil isoforms but insufficient affinity for mouse or rat NK1R. Translationally, this potent antagonism regulates gastric motility and improves the symptoms of motion sickness by inhibiting emetic signaling, while effectively suppressing spontaneous alcohol cravings and blunting stress-induced cortisol responses in clinical settings.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library