Skip to Content
MilliporeSigma

SML4477

Biotin-4-aminophenol, APEX2 Substrate

≥98% (HPLC)

Synonym(s):

(3aS,4S,6aR)-Hexahydro-N-(4-hydroxyphenyl)-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide, BP5, N-(4-hydroxyphenyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H21N3O3S
CAS Number:
Molecular Weight:
335.42
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Biotin-4-aminophenol (BP5) is a high-reactivity, cell-permeable APEX2 substrate engineered to resolve the spatiotemporal dynamics of cytosolic protein complexes with superior specificity. Mechanistically, BP5 generates short-lived phenoxyl radicals upon H2O2 activation that covalently tag unmodified tyrosine residues while preserving phosphorylated sites (pTyr). Crucially, its elevated reactivity dictates a tighter effective capture radius (<10 nm effective capture) via rapid radical quenching and self-polymerization, thereby eliminating the broad "diffusion cloud" and background noise associated with traditional Biotin-Phenol (BP1). This physiochemical refinement prioritizes the labeling of direct interactors over bystanders, enabling the high-fidelity capture of weak and transient assemblies often lost during cell lysis. Validated against highly dynamic signaling networks—including EGFR adaptors (GRB2, STS1) and the ILK complex—BP5 successfully differentiates interactions indistinguishable by BP1, providing a robust platform for mapping mobile interactomes and protein translocation events at organelle interfaces.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library