Skip to Content
MilliporeSigma

SML4478

Difluorocarboxyfluorescein-488 NHS Ester 5-isome

≥95% (HPLC)

Synonym(s):

1-[[(2′,7′-difluoro-3′,6′-dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthen]-5-yl)carbonyl]oxy]-2,5-pyrrolidinedione, 2′,7′-difluoro-3′,6′-dihydroxy-3-oxo-spiro[isobenzofuran-1(3H),9′-[9H]xanthene]-5-carboxylic acid 2,5-dioxo-1-pyrrolidinyl ester, 2-(2,7-Difluoro-6-hydroxy-3-oxo-3H-xanthen-9-yl)-5-(((2,5-dioxopyrrolidin-1-yl)oxy)carbonyl)benzoic acid succinimidyl ester, DFCF-488 NHS Dye

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H13F2NO9
CAS Number:
Molecular Weight:
509.37
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

2‘,7‘-Difluorofluorescein-5-carboxy-NHS ester 488 (DFCF-488, NHS), is an amine-reactive fluorescent dye with a high molar extinction coefficient (ε = 72,000 M⁻¹cm⁻¹), enabling efficient covalent conjugation to antibodies, peptides, proteins, and nucleic acid tracers at pH 7.5-8.5 or lower. With a pKa of ~4.7 and λₑₓ/λₑₘ ~ 490/520 nm, DFCF-488 offers high quantum yield, photostability, and pH-independent fluorescence in physiological conditions, making it ideal for calcium monitoring and pH tracking in acidic organelles and pathways. DFCF-488 conjugates support diverse applications, including live-cell imaging, FRET-based biosensing, high-throughput screening, and immunofluorescence, ensuring reliable performance for cellular labeling and detection.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library