Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C38H62N10O12 · xC2H4O2
Molecular Weight:
850.96 (free base basis)
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
(Powder or Lyophilized powder or Film)
Storage condition:
desiccated
Quality Segment
assay
≥95% (HPLC)
form
(Powder or Lyophilized powder or Film)
storage condition
desiccated
color
white to off-white
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Rytvela (101.10) is a highly cell-permeable, all-D-amino acid heptapeptide engineered to confer absolute protease resistance and structural stability, mimicking the flexible extracellular IL-1RAcP D3-juxtamembranous loop to non-competitively antagonize the interleukin-1 receptor (IL-1R1) complex with exceptional quantitative potency (IC50 ≈ 0.5–2.0 nM; KD = 5 nM). Binding an allosteric pocket distinct from the orthosteric site ensures exquisite IL-1R1 specificity, demonstrated by zero functional efficacy in knockout models and an absence of cross-reactivity with IL-18, IL-6, or TNF-α pathways. By exerting targeted functional selectivity, the reversible biased ligand potently ablates downstream SAPK/JNK, AP-1, and RhoA/ROCK signaling while critically sparing cytoprotective NF-κB activation, thereby circumventing the severe apoptotic toxicity characteristic of global IL-1 blockade induced by indiscriminate orthosteric biologics. Displays superior penetrance across placental, blood-retinal, and epithelial barriers, enabling rytvela to suppress pathogenic Sema3A, MMP9, and CCL2 expression, revert amoeboid microglia to a resting ramified state, prevent endothelial apoptosis, block vascular vaso-obliteration, and abolish pathogenic uterine contractility. Proving highly efficacious whether administered systemically (1 mg/kg IV, SC, IP) or locally (1 mg/kg/day intrarectal; 100 nM topical), rytvela rapidly reverses IL-1-induced hyperthermia, TNBS-induced inflammatory bowel disease, contact dermatitis, OIR-induced retinopathy, and LPS/LTA-induced preterm birth. Ultimately, the novel heptapeptide establishes a translatable, precision-engineered therapeutic paradigm for compartmentalized inflammatory diseases without compromising baseline host defenses.
Disclaimer
Hygroscopic, Store with desiccant
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.