Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C38H46O8
CAS Number:
Molecular Weight:
630.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
Gambogenic acid (GNA) is a promising antitumor agent targeting EZH2 and modulating signaling pathways to inhibit proliferation, metastasis, enhance chemosensitivity, and overcome drug resistance. Acting in a time- and dose-dependent manner, GNA targets cancer hallmarks with broad activity by directly and covalently binding EZH2 (Cys668), disrupting PRC2, activating RIP1/p53/SLC7A11/GPX4, activating GSK3β while inhibiting Akt/mTOR, inducing ferroptosis, and downregulating metastasis proteins. These actions, mediated through pathways like Akt/mTOR, ROS/JNK, and NF-ĸB, result in tumor cell death and proliferation reduction, with GNA showing advantages over gambogic acid in antitumor effect and toxicity. In vitro and in vivo studies confirm GNA′s efficacy, but consideration should be given to potential drug-drug interactions due to GNA′s increase of liver CYP enzyme activity.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
