Skip to Content
MilliporeSigma

SML4493

Carfilzomib

≥98% (HPLC)

Synonym(s):

(aS)-a-[[2-(4-Morpholinyl)acetyl]amino]benzenebutanoyl-L-leucyl-N-[(1S)-3-methyl-1-[[(2R)-2-methyl-2-oxiranyl]carbonyl]butyl]-L-phenylalaninamide, PR 171, PR-171

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C40H57N5O7
CAS Number:
Molecular Weight:
719.91
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
(Powder or Lyophilized powder or Film)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

(Powder or Lyophilized powder or Film)

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Carfilzomib (PR-171) is a definitive, cell-permeable, irreversible tetrapeptide epoxyketone proteasome inhibitor that selectively targets the chymotrypsin-like (ChT-L) activity of the constitutive 20S proteasome (β5) and immunoproteasome (LMP7/β5i) with nanomolar potency (IC50 < 5 nM). By forming specific morpholino adducts with catalytic N-terminal threonines, PR-171 decouples pharmacokinetics from pharmacodynamics, ensuring sustained target suppression despite rapid systemic clearance (t1/2 < 30 min). Unlike boronic acid predecessors, the epoxyketone exhibits negligible affinity for non-proteasomal serine proteases (e.g., cathepsin G, elastase), resulting in a superior neurotoxicity profile. Mechanistically, Carfilzomib blocks NF-κB signaling via IκBα stabilization and drives the accumulation of poly-ubiquitinated proteins, precipitating catastrophic ER stress, mitochondrial depolarization, and JNK-mediated apoptosis. Clinically validated for relapsed/refractory multiple myeloma, PR-171 overcomes bortezomib resistance and demonstrates profound synergy with dexamethasone, lenalidomide and daratumumab in high-risk phenotypes.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library