Skip to Content
MilliporeSigma

STS0023

Greener Alternative Detergents Kit

non-ionic

Sustainability features

Synonym(s):

ECOSURF™ EH-9, TERGITOL™ 15-S-7

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161902
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Sustainability features

application(s)

life science and biopharma

greener alternative category

General description

Surfactants are essential for numerous chemical applications – as detergents, wetting agents, emulsifiers, foaming agents, dispersants and more. Greener Alternative Detergents are now available. This Kit includes various detergents for evaluation in your Research. The included detergents are readily biodegradable (OECD 301). These products are aligned with one or more of “The 12 Principles of Green Chemistry”.

Features and Benefits

The included detergents are readily biodegradable (OECD 301)

Legal Information

ECOSURF is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow
TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • TERGITOL™, Type 15-S-7
    SDS

  • TERGITOL™, Type 15-S-9
    SDS

  • TERGITOL™ 15-S-40 solution, 70% aqueous solution
    SDS

  • ECOSURF™ EH-9 solution, 90% aqueous solution
    SDS

  • TERGITOL™ 15-S-15, Non-ionic surfactant
    SDS

  • ECOSURF™ SA-7, Non-ionic surfactant
    SDS

  • TERGITOL™ 15-S-3, Non-ionic surfactant
    SDS

See All (8)

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library