Skip to Content
MilliporeSigma

STS0029

CHAPS hydrate

BioUltra, ≥98% (HPLC)

Synonym(s):

(3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S · xH2O
Molecular Weight:
614.88 (anhydrous basis)
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Segment

assay

≥98% (HPLC)

General description

CHAPS hydrate, a zwitterionic detergent derived from Cholic acid, stands out as an excellent tool in biochemical and cell biology research, particularly in the solubilization of membrane proteins. This versatile detergent, combining features of sulfobetaine-type and bile-acid detergents, exhibits a remarkable solubility rate of over 50% in aqueous solutions. It serves a pivotal role in stabilizing protein-DNA complexes, ensuring the retention of biochemical activity in solution. Widely utilized as a buffer in various research applications, CHAPS hydrate acts as an amphiphilic surfactant, proving effective in solubilizing plasma membrane proteins and diluting assay components. Notably, its easy removal through dialysis or gel filtration adds to its appeal, making it a preferred choice for molecular mass determination by fluorometry. This feature underscores CHAPS hydrate′s adaptability in diverse experimental setups. However, caution is advised in assays involving polyvalent metals or metal ions sensitive to nonspecific chelating agents. Beyond its conventional use, CHAPS hydrate finds application as a polymer matrix in binding assays, effectively removing metal ions from protein solutions.

Application

0.2 w/v of CHAPS has been used to remove cerebral endothelium by brain perfusion.[1] 2% (w/v) CHAPS has been used as rehydration buffer along with Thiourea and Urea in protein extraction.[2] 10 mM CHAPS has been used along with 0.15 M NaCl, Tris–Cl, pH 7.6, EDTA as lysis buffer in Western Blot analysis.[3] CHAPS has been used as hybridization buffer in in-situ hybridization method of immature mammalian CNS.[4]

Features and Benefits

1. Tested for trace elements or metals that could interfere with experimental results

2. Suitable for Biochemical and Cell Biology research

3. Can be used in Dialysis and Electrophoresis

4. High purity product for research applications


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library